missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thrombospondin-2 Antibody (MM0568-9T19), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-11923B
This item is not returnable.
View return policy
Description
Thrombospondin-2 Monoclonal specifically detects Thrombospondin-2 in Human samples. It is validated for Western Blot.
Specifications
| Thrombospondin-2 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| THBS2 | |
| Human recombinant Thrombospondin-2 | |
| 0.1 mL | |
| 7058 | |
| Store at 4C in the dark. | |
| IgG2 |
| Western Blot | |
| MM0568-9T19 | |
| Western Blot | |
| thrombospondin 2, TSP2thrombospondin-2 | |
| Mouse | |
| Protein G purified | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction