missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Thrombospondin 2 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA547172
This item is not returnable.
View return policy
Description
In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) Thrombospondin-1 and rhThrombospondin-4 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
The Thrombospondin proteins (TSP 1-5) compose a family of glycoproteins that are involved in cell-to-cell and cell-to-matrix signaling. These extracellular, cell-surface proteins form complexes of both homo- and hetero-multimers. Thrombospondins play a role in development, aggregation of platelets, adhesion and migration of cells and progression of cells through the growth cycle. Thrombospondin 1 is released from platelets in response to Thrombin stimulation and is a transient component of the extracellular matrix of developing and repairing tissues. Thrombospondin 2 shares a high degree of homology with Thrombospondin 1, and is thought to have overlapping but unique functions. Thrombospondin 3 is a developmentally regulated heparin-binding protein. Thrombospondin 4 is neuronally expressed and stimulates neurite outgrowth.
Specifications
| Thrombospondin 2 | |
| Polyclonal | |
| Unconjugated | |
| THBS2 | |
| Thbs2; Thbs-2; thrombospondin 2; Thrombospondin2; thrombospondin-2; TSP2; TSP-2; TSP2thrombospondin-2 | |
| Goat | |
| Antigen Affinity Chromatography | |
| RUO | |
| 7058 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| P35442 | |
| THBS2 | |
| Mouse myeloma cell line NS0-derived recombinant human Thrombospondin-2 Gly19-Ile1172. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction