missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thrombospondin-3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94865-0.02ml
This item is not returnable.
View return policy
Description
Thrombospondin-3 Polyclonal antibody specifically detects Thrombospondin-3 in Mouse, Rat samples. It is validated for Western Blot
Specifications
| Thrombospondin-3 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| MGC119565, thrombospondin 3, thrombospondin-3, TSP3MGC119564 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 787-956 of human THBS3 (NP_009043.1). TDDDYAGFLFSYQDSGRFYVVMWKQTEQTYWQATPFRAVAQPGLQLKAVTSVSGPGEHLRNALWHTGHTPDQVRLLWTDPRNVGWRDKTSYRWQLLHRPQVGYIRVKLYEGPQLVADSGVIIDTSMRGGRLGVFCFSQENIIWSNLQYRCNDTVPEDFEPFRRQLLQGRV | |
| 0.02 mL | |
| Cell Biology, Cytoskeleton Markers, Neuroscience, Signal Transduction | |
| 7059 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction