missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thromboxane synthase Antibody (SR1967), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22236-50ul
This item is not returnable.
View return policy
Description
Thromboxane synthase Monoclonal antibody specifically detects Thromboxane synthase in Human, Mouse samples. It is validated for Western Blot
Specifications
| Thromboxane synthase | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| SR1967 | |
| Western Blot 1:500-1:2000 | |
| CYP5A1family 5, subfamily A, polypeptide 1, CYP5FLJ52771, EC 5.3.99.5, GHOSAL, platelet, cytochrome P450, subfamily V, thromboxane A synthase 1 (platelet), thromboxane A synthase 1 (platelet, cytochrome P450, family 5, subfamily A), thromboxane A synthase 1 (platelet, cytochrome P450, subfamily V), thromboxane-A synthase, TXA synthase | |
| A synthesized peptide derived from human Thromboxane synthase (Uniprot #: P24557) | |
| 50 μg | |
| Lipid and Metabolism | |
| 6916 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction