missing translation for 'onlineSavingsMsg'
Learn More

THRSP Antibody, Allophycocyanin, Novus Biologicals™

Product Code. 18679891 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18679891 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18679891 Supplier Novus Biologicals Supplier No. NBP298508APC

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

THRSP Polyclonal antibody specifically detects THRSP in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen THRSP
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate APC
Formulation PBS
Gene Alias lipogenic protein 1, Lpgp, LPGP1, S14MGC21659, Spot 14 protein, SPOT14SPOT14 homolog, thyroid hormone responsive, thyroid hormone responsive (SPOT14 homolog, rat), thyroid hormone responsive SPOT14, thyroid hormone responsive SPOT14 (rat) homolog, thyroid hormone-inducible hepatic protein
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human THRSP fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 7069
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.