missing translation for 'onlineSavingsMsg'
Learn More
Learn More
THSD7A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
€ 285.00 - € 549.00
Specifications
| Antigen | THSD7A |
|---|---|
| Concentration | 0.1mg/mL |
| Dilution | Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18495251
|
Novus Biologicals
NBP1-93612-25ul |
25 μL |
€ 285.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18036437
|
Novus Biologicals
NBP1-93612 |
0.1 mL |
€ 549.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
THSD7A Polyclonal specifically detects THSD7A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| THSD7A | |
| Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| thrombospondin, type I, domain containing 7A | |
| THSD7A | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| 0.1mg/mL | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 221981 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:VCQSSPCEAEELRYSLHVGPWSTCSMPHSRQVRQARRRGKNKEREKDRSKGVKDPEARELIKKKRNRNRQNRQENKYWDIQIGYQTREVMCINKTGKAADLSFCQQEKLPMTFQSC | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title