missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ THSD7B Polyclonal Antibody
GREENER_CHOICE

Product Code. 15710202 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15710202 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15710202 Supplier Invitrogen™ Supplier No. PA562404

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: VLMESTGPAG HCPHLVESVP CEDPMCYRWL ASEGICFPDH GKCGLGHRIL KAVCQNDRGE DVSGSLCPVP PPPERKSCEI PCRMDCVLSE W Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 90%.

THSD7B is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen THSD7B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene THSD7B
Gene Accession No. Q9C0I4
Gene Alias KIAA1679; thrombospondin type 1 domain containing 7B; thrombospondin type-1 domain-containing protein 7B; thrombospondin, type I, domain containing 7B; THSD7B
Gene Symbols THSD7B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human THSD7B. Recombinant protein control fragment (Product #RP-101354).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80731
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.