missing translation for 'onlineSavingsMsg'
Learn More
Learn More
THTPA Antibody (OTI13E3), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74511AF700
This item is not returnable.
View return policy
Description
THTPA Monoclonal specifically detects THTPA in Human samples. It is validated for Western Blot.
Specifications
| THTPA | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| THTPA | |
| Recombinant protein fragment corresponding to amino acids 2-230 of human THTPA (NP_077304) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2b |
| Western Blot | |
| OTI13E3 | |
| Western Blot | |
| EC 3.6.1.28, MGC2652, thiamine triphosphatase, thiamine-triphosphatase, THTP, THTPASE | |
| Mouse | |
| Affinity Purified | |
| Lipid and Metabolism | |
| 79178.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction