missing translation for 'onlineSavingsMsg'
Learn More
Learn More
THTPA Monoclonal Antibody (OTI4H2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA806044
This item is not returnable.
View return policy
Description
THTPA encodes an enzyme which catalyzes the biosynthesis of thiamine disphophate (vitamin B1) by hydrolysis of thiamine triphosphate. Alternative splicing results in multiple transcript variants.Specifications
| THTPA | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9BU02 | |
| THTPA | |
| Human recombit protein fragment corresponding to amino acids 2-230 of human THTPA produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot | |
| OTI4H2 | |
| Unconjugated | |
| THTPA | |
| THTP, THTPASE | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 79178 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction