missing translation for 'onlineSavingsMsg'
Learn More
Learn More
THUMPD3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93482-0.02ml
This item is not returnable.
View return policy
Description
THUMPD3 Polyclonal antibody specifically detects THUMPD3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| THUMPD3 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| DKFZP434F091, THUMP domain containing 3, THUMP domain-containing protein 3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-250 of human THUMPD3 (NP_056268.2). MCDIEEATNQLLDVNLHENQKSVQVTESDLGSESELLVTIGATVPTGFEQTAADEVREKLGSSCKISRDRGKIYFVISVESLAQVHCLRSVDNLFVVVQEFQDYQFKQTKEEVLKDFEDLAGKLPWSNPLKVWKINASFKKKKAKRKKINQNSSKEKINNGQEVKIDQRNVKKEFTSHALDSHILDYYENPAIKEDVSTLIGDDLASCKDETDESSKEETEPQVLKFRVTCNRAGEKHCFTSNEAARDFG | |
| 0.02 mL | |
| Cell Biology | |
| 25917 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction