missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thymidine Kinase 1 Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-60644V
This item is not returnable.
View return policy
Description
Thymidine Kinase 1 Polyclonal antibody specifically detects Thymidine Kinase 1 in Human samples. It is validated for Western Blot
Specifications
| Thymidine Kinase 1 | |
| Polyclonal | |
| Western Blot | |
| EC 2.7.1.21, thymidine kinase 1 soluble isoform, thymidine kinase 1, soluble, thymidine kinase, cytosolic, thymidine kinase-1, TK2 | |
| The immunogen recognized by this antibody maps to a region between residue 205 and 234 of human thymidine kinase 1 using the numbering given in entry NP_003249.1 (GeneID 7083). | |
| 0.1 mL | |
| DNA Repair, DNA replication Transcription Translation and Splicing, Stem Cell Markers | |
| 7083 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction