missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thymidylate Synthase Antibody (TYMS/9091R), DyLight 550, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-24023R
This item is not returnable.
View return policy
Description
Thymidylate Synthase Monoclonal antibody specifically detects Thymidylate Synthase in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Thymidylate Synthase | |
| Monoclonal | |
| DyLight 550 | |
| EC 2.1.1.45, HsT422, MGC88736, thymidylate synthase, thymidylate synthetase, TMSTsase, TSase, TSHST422 | |
| Recombinant human Thymidylate Synthase protein fragment (around aa 1-200) (exact sequence is proprietary) | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, DNA Repair, DNA replication Transcription Translation and Splicing, Transcription Factors and Regulators, Translation Control | |
| 7298 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| TYMS/9091R | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction