missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Thymol Blue
Click to view available options
Quantity:
10 g
50 g
Description
A sulfonphthalein dye used as a pH indicator.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
A sulfonphthalein dye used as a pH indicator.
Solubility
Solubility in 0.1 N sodium hydroxide solution, almost transparent. Soluble in alcohol, dilute alkali, 0.1% ethanol. Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
A sulfonphthalein dye used as a pH indicator.
Solubility
Solubility in 0.1 N sodium hydroxide solution, almost transparent. Soluble in alcohol, dilute alkali, 0.1% ethanol. Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Melting Point | 223°C (decomposition) |
Merck Index | 14,9400 |
Formula Weight | 466.6 |
Quantity | 10 g |
Chemical Name or Material | Thymol Blue |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'