Learn More
Invitrogen™ THYN1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114042
Description
Immunogen sequence: NPHYDPSSKE DNPKWSMVDV QFVRMMKRFI PLAELKSYHQ AHKATGGPLK For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 86%; Dog: 93%; Guinea Pig: 92%; Horse: 100%; Human: 100%; Mouse: 93%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 92%.
THYN1, also named as THY28, is a 28-32 kDa protein which located mainly in the nucleus. It appears to correlate with induction of apoptosis.
Specifications
| THYN1 | |
| Polyclonal | |
| Unconjugated | |
| THYN1 | |
| D730042P09Rik; HSPC144; MDS012; mThy28; My0054; MY105; Thy28; THY28KD; Thymocyte nuclear protein 1; thymocyte protein thy28; Thyn1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 29087 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9P016 | |
| THYN1 | |
| Synthetic peptide directed towards the middle region of human THYN1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.