Learn More
Thyroglobulin Antibody (CL0164) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-44463
Product Code. 30287774
  • € 561.00 / 100µL
Description
Thyroglobulin Monoclonal antibody specifically detects Thyroglobulin in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin),Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Thyroglobulin
Monoclonal
LYOPH
Western Blot 1 μg/mL, Immunohistochemistry-Paraffin 1:500 - 1:1000
AITD3TGN, TDH3, Tg, thyroglobulin
This antibody was generated using a recombinant protein sequence of P01266, with the exact immunogen sequence remaining proprietary.
100 μL
Cancer, Cell Biology
7038
Human
Purified
Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry
CL0164
Unconjugated
Lyophilized in 5% Trehalose.
Mouse
Protein A purified
RUO
Primary
Centrifuge the vial of lyophilized antibody at 12,000 x g for 20 seconds. Reconstitute by adding sterile, distilled water to achieve a final antibody concentration of 1mg/mL.
Store at 4⪚C short term. Aliquot and store at -20⪚C long term. Avoid freeze-thaw cycles.
IgG2b