missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thyroglobulin Antibody (rTGB/4744), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08264B
This item is not returnable.
View return policy
Description
Thyroglobulin Monoclonal antibody specifically detects Thyroglobulin in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin)
Specifications
| Thyroglobulin | |
| Monoclonal | |
| Biotin | |
| AITD3TGN, TDH3, Tg, thyroglobulin | |
| Human thyroid follicular cells | |
| 100 μg | |
| Cancer, Cell Biology | |
| 7038 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Flow Cytometry, Immunohistochemistry (Paraffin) | |
| rTGB/4744 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction