missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thyroglobulin Antibody (TGB24), DyLight 350 , Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-11600UV
This item is not returnable.
View return policy
Description
Thyroglobulin Monoclonal antibody specifically detects Thyroglobulin in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Thyroglobulin | |
| Monoclonal | |
| DyLight 350 | |
| AITD3TGN, TDH3, Tg, thyroglobulin | |
| Human thyroid follicular cells | |
| 0.1 mL | |
| Cancer, Cell Biology | |
| 7038 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| TGB24 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction