missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thyroglobulin Mouse anti-Human, Clone: 4D3C5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60272-1-IG-20UL
This item is not returnable.
View return policy
Description
Thyroglobulin(Tg) is a glycoprotein homodimer produced predomitly by the thryroid gland. It acts as a substrate for the synthesis of thyroxine and triiodothyronine as well as the storage of the inactive forms of thyroid hormone and iodine.Thyroglobulinis secreted from the endoplasmic reticulum to its site of iodination, and subsequent thyroxine biosynthesis, in the follicular lumen. Mutations in this gene cause thyroid dyshormonogenesis, manifested as goiter, and are associated with moderate to severe congenital hypothyroidism. Polymorphisms in this gene are associated with susceptibility to autoimmune thyroid diseases (AITD) such as Graves disease and Hashimoto thryoiditis.Specifications
| Thyroglobulin | |
| Monoclonal | |
| 1.63 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P01266 | |
| TG | |
| Thyroglobulin Fusion Protein Ag18070 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| 4D3C5 | |
| Unconjugated | |
| TG | |
| AITD3, TG, TGN, thyroglobulin | |
| Mouse | |
| Protein A | |
| RUO | |
| 7038 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction