Learn More
Thyroglobulin Mouse anti-Human, CoraLite 594, Clone: 4D3C5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL594-60272-100UL
Description
The Thyroglobulin antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Thyroglobulin. This antibody recognizes human antigen. The Thyroglobulin antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Specifications
| Thyroglobulin | |
| Monoclonal | |
| CoraLite 594 | |
| 100 μL | |
| 7038 | |
| IgG2a |
| Immunofluorescence | |
| 4D3C5 | |
| Mouse | |
| Primary | |
| Human |