Learn More
Thyroglobulin Mouse anti-Human, CoraLite 594, Clone: 4D3C5, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL594-60272-100UL
Product Code. 17412653
  • € 450.00 / 100µL
Description
The Thyroglobulin antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Thyroglobulin. This antibody recognizes human antigen. The Thyroglobulin antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Specifications
Specifications
Thyroglobulin
Monoclonal
CoraLite 594
100 μL
7038
IgG2a
Immunofluorescence
4D3C5