missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIA1 Antibody (TIA1/1313), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54406PE
This item is not returnable.
View return policy
Description
TIA1 Monoclonal antibody specifically detects TIA1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Flow (Intracellular), CyTOF
Specifications
| TIA1 | |
| Monoclonal | |
| PE | |
| cytotoxic granule-associated RNA-binding protein, nucleolysin TIA-1 isoform p40, p40-TIA-1, p40-TIA-1 (containing p15-TIA-1), RNA-binding protein TIA-1, T-cell-restricted intracellular antigen-1, TIA-1, TIA1 cytotoxic granule-associated RNA binding protein, TIA1 cytotoxic granule-associated RNA-binding protein | |
| Recombinant human TIA1 fragment of 102 amino acid residues (aa279-380) (Uniprot: P31483) | |
| 100 μg | |
| Adaptive Immunity, Apoptosis, Immunology | |
| 7072 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Western Blot, Flow Cytometry, Immunofluorescence, Flow Cytometry (Intracellular Staining), CyTOF | |
| TIA1/1313 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction