missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SP110 Mouse anti-Human, Clone: OTI4C1, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA807854S
This item is not returnable.
View return policy
Description
SP110 Monoclonal antibody specifically detects SP110 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| SP110 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9HB58 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 3431 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4C1 | |
| Unconjugated | |
| SP110 | |
| 5031415C07Rik; 52kDa; 5830484A20Rik; ENSMUSG00000075603; IFI41; Ifi75; interferon-induced protein 41, 30kD; interferon-induced protein 41/75; interferon-induced protein 75, 52kD; intracellular pathogen resistance 1; intracellular pathogen resistance protein 1; Ipr1; phosphoprotein 41; phosphoprotein 75; SP110; Sp110 nuclear body protein; speckled 110 kDa; Transcriptional coactivator Sp110; VODI | |
| Human recombinant protein fragment corresponding to amino acids 232-521 of human SP110 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction