missing translation for 'onlineSavingsMsg'
Learn More

TIA1 Mouse anti-Human, Clone: TIA1/1313, Abnova™

Product Code. 16093206
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16093206 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16093206 Supplier Abnova Supplier No. MAB14898.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against partial recombinant human TIA1.

  • High specificity
  • High reactivity

Specifications

Antigen TIA1
Applications Flow Cytometry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone TIA1/1313
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant human TIA1.
Dilution Flow Cytometry (0.5-1 ug/million cells) Immunofluorescence (1-2 ug/mL) Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 10 mM PBS.
Gene Symbols TIA1
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 279-380 of human TIA1 (exact sequence is proprietary).
Purification Method Protein A purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7072
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.