missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIAL1 Antibody (6E3), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-11834MFV610
This item is not returnable.
View return policy
Description
TIAL1 Monoclonal antibody specifically detects TIAL1 in Human, Mouse, Primate samples. It is validated for Flow Cytometry, Flow (Intracellular)
Specifications
| TIAL1 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Primate | |
| Purified |
| Flow Cytometry | |
| 6000 | |
| Flow Cytometry, Flow (Intracellular) | |
| aging-associated gene 7 protein, MGC33401, TCBP, T-cluster binding protein, TIA1 cytotoxic granule-associated RNA binding protein-like 1, TIA1 cytotoxic granule-associated RNA-binding protein-like 1, TIA-1 related protein, TIA-1-related nucleolysin, TIA-1-related protein, TIARnucleolysin TIAR | |
| GST-tagged recombinant human TIAL1 | |
| 0.1 mL | |
| Apoptosis | |
| 7073 | |
| Store at 4C in the dark. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction