missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TIAL1 Monoclonal Antibody (6E3), Alexa Fluor™ 488

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA544139
This item is not returnable.
View return policy
Description
This antibody recognizes TIAR, an ubiquitously expressed approximately 42 kDa intracellular RNA-binding protein.
The protein encoded by this gene is a member of a family of RNA-binding proteins, has three RNA recognition motifs (RRMs), and binds adenine and uridine-rich elements in mRNA and pre-mRNAs of a wide range of genes. It regulates various activities including translational control, splicing and apoptosis. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. The different isoforms have been show to function differently with respect to post-transcriptional silencing.
Specifications
| TIAL1 | |
| Monoclonal | |
| 0.1 mg/mL | |
| PBS with 0.2% BSA and 15mM sodium azide | |
| P70318, Q01085 | |
| TIAL1 | |
| GST-tagged recombinant human TIAR. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Monkey | |
| Antibody | |
| IgG2a κ |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 6E3 | |
| Alexa Fluor 488 | |
| TIAL1 | |
| 5330433G13Rik; aging-associated gene 7 protein; AL033329; mTIAR; Nucleolysin TIAR; RNA binding protein TIAR (TIA-1 related); TCBP; T-cluster binding protein; Tia1 cytotoxic granule-associated RNA binding protein-like 1; TIA-1-related nucleolysin; TIA-1-related protein; Tial 1 cytotoxic granule-associated RNA- binding protein-like 1; TIAL1; Tial1 cytotoxic granule-associated RNA binding protein-like 1; Tial1 cytotoxic granule-associated RNA-binding protein-like 1; TIAR | |
| Mouse | |
| Size-exclusion chromatography | |
| RUO | |
| 21843, 7073 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction