missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TIAM2 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142639
This item is not returnable.
View return policy
Description
Additional Information: This antibody is expected to recognize isoform a (NP_036586.2) only. This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:64,000.
Modulates the activity of RHO-like proteins and connects extracellular signals to cytoskeletal activities. Acts as a GDP- dissociation stimulator protein that stimulates the GDP-GTP exchange activity of RHO-like GTPases and activates them. Mediates extracellular laminin signals to activate Rac1, contributing to neurite growth. Involved in lamellipodial formation and advancement of the growth cone of embryonic hippocampal neurons. Promotes migration of neurons in the cerebral cortex. When overexpressed, induces membrane ruffling accompanied by the accumulation of actin filaments along the altered plasma membrane. Activates specifically RAC1, but not CDC42 and RHOA.
Specifications
| TIAM2 | |
| Polyclonal | |
| Unconjugated | |
| Tiam2 | |
| 3000002F19Rik; KIAA2016; mKIAA2016; RAS-related C3 botulinum substrate 1, guanine nucleotide exchange factor 1; SIF and TIAM1-like exchange factor; Stef; T cell lymphoma invasion and metastasis 2; T-cell lymphoma invasion and metastasis 2; T-cell lymphoma invasion and metastasis 2; RAS-related C3 botulinum substrate 1, guanine nucleotide exchange factor 1; TIAM2; TIAM-2; T-lymphoma invasion and metastasis-inducing protein 2 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 26230 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q8IVF5 | |
| Tiam2 | |
| Peptide with sequence TLQGSKNHSNT-C (aa 9-19). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction