missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tiapride EP Impurity B, Mikromol™
Description
Tiapride EP Impurity B
Specifications
Specifications
| Chemical Name or Material | 2-Methoxy-5-(methylsulfonyl)benzoic Acid |
| CAS | 50390-76-6 |
| Analyte or Component Names | 2-Methoxy-5-(methylsulfonyl)benzoic Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C9 H10 O5 S |
| Certifications/Compliance | ISO 17025 |
| Synonym | 2-Methoxy-5-(methylsulfonyl)benzoic acid, 5-Methylsulfonyl-2-methoxybenzoic acid, 6-Methoxy-3-methylsulfonylbenzoic acid, 5-mesyl-o-anisic acid, Tiapride Hydrochloride Impurity B (EP), Tiapride Hydrochloride Imp. B (EP), Tiapride Imp. B (EP), Tiapride Hydrochloride Impurity B, Tiapride Impurity B |
| InChI Formula | InChI=1S/C9H10O5S/c1-14-8-4-3-6(15(2,12)13)5-7(8)9(10)11/h3-5H,1-2H3,(H,10,11) |
| SMILES | COc1ccc(cc1C(=O)O)S(=O)(=O)C |
| IUPAC Name | 2-methoxy-5-methylsulfonylbenzoic acid |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?