missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tiaprofenic Acid 1,3-Butylene Glycol Esters (Mixture of Regio- and Stereoisomers), Mikromol™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Tiaprofenic Acid 1,3-Butylene Glycol Esters (Mixture of Regio- and Stereoisomers)
Specifications
Specifications
| Chemical Name or Material | Tiaprofenic Acid 1,3-Butylene Glycol Esters (Mixture of Regio- and Stereoisomers) |
| Analyte or Component Names | Tiaprofenic Acid 1,3-Butylene Glycol Esters (Mixture of Regio- and Stereoisomers) |
| Shipping Condition | Room Temperature |
| Molecular Formula | 2 C18 H20 O4 S |
| Certifications/Compliance | ISO 9001 |
| Synonym | Trifluoperazine N1,N4,S-Trioxide, Tiaprofenic Acid 1,3-Butylene Glycol Ester (Mixture of Isomers) |
| InChI Formula | InChI=1S/2C18H20O4S/c1-12(19)10-11-22-18(21)13(2)15-8-9-16(23-15)17(20)14-6-4-3-5-7-14;1-12(10-11-19)22-18(21)13(2)15-8-9-16(23-15)17(20)14-6-4-3-5-7-14/h2*3-9,12-13,19H,10-11H2,1-2H3 |
| SMILES | CC(O)CCOC(=O)C(C)c1ccc(s1)C(=O)c2ccccc2.CC(CCO)OC(=O)C(C)c3ccc(s3)C(=O)c4ccccc4 |
| IUPAC Name | 4-hydroxybutan-2-yl 2-(5-benzoylthiophen-2-yl)propanoate;3-hydroxybutyl 2-(5-benzoylthiophen-2-yl)propanoate |
| Molecular Weight (g/mol) | 664.83 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction