missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Tibulizumab Humanized Recombinant Human Monoclonal Antibody

Human Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA542177
This item is not returnable.
View return policy
Description
Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Tibulizumab is a tetravalent bispecific antibody targeting BAFF and IL-17A for the treatment of autoimmune disease.
Specifications
| Tibulizumab Humanized | |
| Recombinant Monoclonal | |
| Unconjugated | |
| LY 3090106; LY3090106; LY-3090106 | |
| Human IL17A/TNFSF13B/CD257. | |
| 100 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| Chemical |
| ELISA | |
| 0.71 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Human | |
| Protein A/G | |
| RUO | |
| Human | |
| Antibody | |
| IgG4 κ |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering