missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Tibulizumab Humanized Recombinant Human Monoclonal Antibody

Human Recombinant Monoclonal Antibody
€ 610.00 - € 2530.00
Specifications
| Antigen | Tibulizumab Humanized |
|---|---|
| Concentration | 1 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30283118
![]() |
Invitrogen™
MA560135 |
100 μg |
€ 610.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30282838
|
Invitrogen™
MA560136 |
1 mg |
€ 2530.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Tibulizumab is a tetravalent bispecific antibody targeting BAFF and IL-17A for the treatment of autoimmune disease.Specifications
| Tibulizumab Humanized | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Lyophilized | |
| IgG4 | |
| Human | |
| LY 3090106; LY3090106; LY-3090106 | |
| Antibody |
| 1 mg/mL | |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| Unconjugated | |
| Human | |
| RUO | |
| 25 mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Primary | |
| Protein A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title