missing translation for 'onlineSavingsMsg'
Learn More

Bandelin Electronic™ Tickopur R 27 Special Acid Cleaners

Product Code. 10323941
Click to view available options
Volume (Metric):
2 L
5 L
Unit Size:
2 L
5 L
This item is not returnable. View return policy

Product Code. 10323941

Brand: Bandelin Electronic™ 816

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Product Type Special Acid Cleaner
Form Liquid
pH 1.9
Product Line Tickopur
Appearance Clear Pale Yellow
Container Type Polyethylene Canister
For Use With (Application) For Cleaning of fittings, water meters, workpieces of industry, handicrafts and Laboratory
Quantity 1 Ea.
Volume (Metric) 5 L
compliance-icons
Product Identifier
  • Tickopur R27
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : phosphoric acid, FATTY ALCOHOL C12-C14 ethoxylated
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.