Learn More
Bandelin Electronic™ Tickopur R 3 Special Acid Cleaners
Product Code. 10481393
Change view
Click to view available options
Volume (Metric):
2 L
5 L
Unit Size:
2 L
5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
10481393 2 L 2 L
10300812 5 L 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10481393 Supplier Bandelin Electronic™ Supplier No. 923
Edge
Specifications
Product Type Special Acid Cleaner
Form Liquid
pH 3
Product Line Tickopur
Appearance Colorless
Container Type Polyethylene Henkel bottle
For Use With (Application) For cleaning, rust removal, decalcification and degreasing of disassembled and undivided Machinery, tools, equipment
Quantity 1 Ea.
Volume (Metric) 2 L