Learn More
Bandelin Electronic™ Tickopur R 33 Neutral Cleaners
Product Code. 10211911
Click to view available options
Volume (Metric):
2 L
5 L
Unit Size:
2 L
5 L
Product Code. 10211911
Brand: Bandelin Electronic™ 811
Specifications
Product Type Neutral Cleaner
Form Liquid
pH 7
Product Line Tickopur
Appearance Clear Pale Yellow
Container Type Polyethylene Canister
For Use With (Application) Metal and light Metal, glass, ceramics, plastics, Rubber
Quantity 1 Ea.
Volume (Metric) 5 L
compliance-icons
Product Identifier
  • Tickopur R30
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : C16-C18 alkanesulfonate, FATTY ALCOHOL C12-C14 ethoxylated,1, 2, 3-Benzotriazol