Learn More
Bandelin Electronic™ Tickopur RW 77 Special Cleaners
Product Code. 10758341
Click to view available options
Volume (Metric):
5 L
Unit Size:
5 L
Product Code. 10758341
Brand: Bandelin Electronic™ 871
Specifications
Product Type Special Alkaline Cleaner
Form Liquid
pH 9.9
Product Line Tickopur
Appearance Clear Pale Yellow
Container Type Polyethylene Canister
For Use With (Application) For Cleaning of disassembled and undivided Machinery and tool parts, printed circuit boards in the service area, clocks, test sieves, coins
Quantity 1 Ea.
Volume (Metric) 5 L
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : C16-C18 Fatty acid TEA, 2-propanol, C13-C17 alkansulfonate,FATTY ALCOHOL C12-C14 ethoxylated