Learn More
Bandelin Electronic™ Tickopur RW 77 Special Cleaners
Click to view available options
Volume (Metric):
5 L
Unit Size:
5 L
Specifications
Specifications
| Product Type | Special Alkaline Cleaner |
| Form | Liquid |
| pH | 9.9 |
| Product Line | Tickopur |
| Appearance | Clear Pale Yellow |
| Container Type | Polyethylene Canister |
| For Use With (Application) | For Cleaning of disassembled and undivided Machinery and tool parts, printed circuit boards in the service area, clocks, test sieves, coins |
| Quantity | 1 Ea. |
| Volume (Metric) | 5 L |
Safety and Handling
Signal Word
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : C16-C18 Fatty acid TEA, 2-propanol, C13-C17 alkansulfonate,FATTY ALCOHOL C12-C14 ethoxylated