missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TID1 Polyclonal Antibody

TID1 Polyclonal Antibody
Brand: Invitrogen PA598660
This item is not returnable.
View return policy
Description
TID1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), IP, ELISA
Specifications
| TID1 | |
| Polyclonal | |
| Unconjugated | |
| DNAJA3 | |
| 1200003J13Rik; 1810053A11Rik; C81173; DnaJ (Hsp40) homolog, subfamily A, member 3; DnaJ heat shock protein family (Hsp40) member A3; dnaJ homolog subfamily A member 3, mitochondrial; DnaJ protein Tid-1; Dnaja3; HCA57; Hepatocellular carcinoma-associated antigen 57; hTID-1; mTid-1; Tid1; Tid-1; Tid1l; Tumorous imaginal discs protein Tid56 homolog | |
| Rabbit | |
| Protein G | |
| RUO | |
| 9093 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 3.64 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q96EY1 | |
| DNAJA3 | |
| Recombinant Human DnaJ homolog subfamily A member 3, mitochondrial protein (86-184AA ). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction