missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tie-1 Antibody (6F12), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-18610AF350
This item is not returnable.
View return policy
Description
Tie-1 Monoclonal specifically detects Tie-1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications
| Tie-1 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| TIE1 | |
| Recombinant human soluble extracellular TIE-1 | |
| 0.1 mL | |
| Primary | |
| The monoclonal antibody will detect native human TIE-1 in ELISA experiments and on the surface of different human cell types. | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, ELISA | |
| 6F12 | |
| Western Blot, Flow Cytometry, ELISA | |
| EC 2.7.10, EC 2.7.10.1, JTK14soluble TIE1 variant 3, receptor tyrosine kinase, soluble TIE1 variant 1, soluble TIE1 variant 2, soluble TIE1 variant 5, TIEsoluble TIE1 variant 4, tyrosine kinase with immunoglobulin and epidermal growth factor homologydomains 1, tyrosine kinase with immunoglobulin-like and EGF-like domains 1, tyrosine-protein kinase receptor Tie-1 | |
| Mouse | |
| Protein G purified | |
| Angiogenesis, Cancer, Protein Kinase | |
| 7075.0 | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction