Learn More
Tie-2 Antibody (83715), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB3131F
Product Code. 18696753
  • € 706.00 / 0.10mL
Description
Tie-2 Monoclonal antibody specifically detects Tie-2 in Human samples. It is validated for Western Blot, Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Tie-2
Monoclonal
FITC
PBS
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Human
Purified
Western Blot, Flow Cytometry
83715
Western Blot, Flow Cytometry
angiopoietin-1 receptor, CD202B, CD202b antigen, EC 2.7.10, EC 2.7.10.1, hTIE2, p140 TEK, soluble TIE2 variant 1, soluble TIE2 variant 2, TEK tyrosine kinase, endothelial, TIE-2, TIE2CD202b, Tunica interna endothelial cell kinase, Tyrosine-protein kinase receptor TEK, Tyrosine-protein kinase receptor TIE-2, venous malformations, multiple cutaneous and mucosal, VMCM, VMCM1
Mouse myeloma cell line NS0-derived recombinant human Tie-2, Ala23-Lys745, Accession # AAA61139
0.1 mL
Angiogenesis, Cancer, Protein Kinase
7010
Store at 4°C in the dark.