missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tie-2 Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-69753FR
This item is not returnable.
View return policy
Description
Tie-2 Polyclonal antibody specifically detects Tie-2 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Tie-2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 680 | |
| angiopoietin-1 receptor, CD202B, CD202b antigen, EC 2.7.10, EC 2.7.10.1, hTIE2, p140 TEK, soluble TIE2 variant 1, soluble TIE2 variant 2, TEK tyrosine kinase, endothelial, TIE-2, TIE2CD202b, Tunica interna endothelial cell kinase, Tyrosine-protein kinase receptor TEK, Tyrosine-protein kinase receptor TIE-2, venous malformations, multiple cutaneous and mucosal, VMCM, VMCM1 | |
| A genomic peptide made to an internal region of the human TIE2 protein (within residues 250-400). [Swiss-Prot Q02763] | |
| 0.1 mL | |
| Angiogenesis, Cancer, Protein Kinase | |
| 7010 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction