missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tie-2 Antibody (Regeneron patent anti-TIE-2), Alexa Fluor™ 405, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP3-27984AF405
This item is not returnable.
View return policy
Description
Tie-2 Monoclonal antibody specifically detects Tie-2 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Tie-2 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| angiopoietin-1 receptor, CD202B, CD202b antigen, EC 2.7.10, EC 2.7.10.1, hTIE2, p140 TEK, soluble TIE2 variant 1, soluble TIE2 variant 2, TEK tyrosine kinase, endothelial, TIE-2, TIE2CD202b, Tunica interna endothelial cell kinase, Tyrosine-protein kinase receptor TEK, Tyrosine-protein kinase receptor TIE-2, venous malformations, multiple cutaneous and mucosal, VMCM, VMCM1 | |
| TIE2/ CD202b | |
| 0.1 mL | |
| Angiogenesis, Cancer, Phospho Specific, Protein Kinase | |
| 7010 | |
| Store at 4°C in the dark. | |
| IgG4 |
| Flow Cytometry, ELISA, Functional Assay | |
| Regeneron patent anti-TIE-2 | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction