Learn More
Tie-2 Antibody (Regeneron patent anti-TIE-2), mFluor Violet 610 SE, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals NBP3-27984MFV610
Product Code. 30116297
  • € 475.00 / 0.10mL
Description
Tie-2 Monoclonal antibody specifically detects Tie-2 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Tie-2
Monoclonal
mFluor Violet 610 SE
angiopoietin-1 receptor, CD202B, CD202b antigen, EC 2.7.10, EC 2.7.10.1, hTIE2, p140 TEK, soluble TIE2 variant 1, soluble TIE2 variant 2, TEK tyrosine kinase, endothelial, TIE-2, TIE2CD202b, Tunica interna endothelial cell kinase, Tyrosine-protein kinase receptor TEK, Tyrosine-protein kinase receptor TIE-2, venous malformations, multiple cutaneous and mucosal, VMCM, VMCM1
TIE2/ CD202b
0.1 mL
Angiogenesis, Cancer, Phospho Specific, Protein Kinase
7010
Store at 4°C in the dark.
IgG4
Flow Cytometry, ELISA, Functional Assay
Regeneron patent anti-TIE-2
50mM Sodium Borate
Human
Protein A purified