missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIF1 alpha Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41220AF750
This item is not returnable.
View return policy
Description
TIF1 alpha Polyclonal antibody specifically detects TIF1 alpha in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| TIF1 alpha | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| Alexa Fluor 750 | |
| E3 ubiquitin-protein ligase TRIM24, EC 6.3.2, EC 6.3.2.-, hTIF1, PTC6, RING finger protein 82, RNF82Tif1a, TIF1-alpha, TIF1ATIF1ALPHA, TIF1TF1A, transcription intermediary factor 1-alpha, transcriptional intermediary factor 1, tripartite motif containing 24, tripartite motif-containing 24, Tripartite motif-containing protein 24 | |
| Antibody was raised against a 19 amino acid peptide near the center of human TRIM24. The immunogen is located within amino acids 530 - 580 of TRIM24. Amino Acid Squence: LRYPPNQNIPRQAIKPNPL | |
| 0.1 mL | |
| Protein Kinase, Zinc Finger | |
| 8805 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction