missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC25A25 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-59870
This item is not returnable.
View return policy
Description
SLC25A25 Polyclonal specifically detects SLC25A25 in Human samples. It is validated for Western Blot.
Specifications
| SLC25A25 | |
| Polyclonal | |
| Unconjugated | |
| PBS and 2% Sucrose with 0.09% Sodium Azide | |
| APC3, calcium-binding mitochondrial carrier protein SCaMC-2, KIAA1896RP11-395P17.4, MCSC, MCSC3, MGC105138, MGC119514, MGC119515, MGC119516, MGC119517, Mitochondrial ATP-Mg/Pi carrier protein 3, Mitochondrial Ca(2+)-dependent solute carrier protein 3, mitochondrial Ca2+-dependent solute carrier, PCSCL, SCAMC2, SCAMC-2, short calcium-binding mitochondrial carrier 2, small calcium-binding mitochondrial carrier 2, Small calcium-binding mitochondrial carrier protein 2, solute carrier family 25 (mitochondrial carrier; phosphate carrier), member 25, Solute carrier family 25 member 25, solute carrier family 25, member 25 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 114789 | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q6KCM6 | |
| SLC25A25 | |
| Synthetic peptides corresponding to SLC25A25(solute carrier family 25 (mitochondrial carrier, phosphate carrier, member 25). The peptide sequence was selected from the N-terminal of SLC25A25. Peptide sequence AVGLRRLGLHRTEGELQKIVQAGDKDLDGQLDFEEFVHYLQDHEKKLRLV The peptide sequence for this immunogen was taken from within the described region. | |
| 100 μL | |
| Primary | |
| Centrifuge prior to opening. Reconstitute with sterilized PBS to a final concentration of 1mg/ml. Vortex followed by Centrifuge again to pellet the solution. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction