missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIF1 alpha Antibody (OTI2D9), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74514JF549
This item is not returnable.
View return policy
Description
TIF1 alpha Monoclonal specifically detects TIF1 alpha in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| TIF1 alpha | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| TRIM24 | |
| Recombinant protein fragment corresponding to amino acids 706-1016 of human TIF1 alpha (NP_003843) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI2D9 | |
| Western Blot, Immunohistochemistry | |
| E3 ubiquitin-protein ligase TRIM24, EC 6.3.2, EC 6.3.2.-, hTIF1, PTC6, RING finger protein 82, RNF82Tif1a, TIF1-alpha, TIF1ATIF1ALPHA, TIF1TF1A, transcription intermediary factor 1-alpha, transcriptional intermediary factor 1, tripartite motif containing 24, tripartite motif-containing 24, Tripartite motif-containing protein 24 | |
| Mouse | |
| Affinity Purified | |
| Protein Kinase, Zinc Finger | |
| 8805.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction