Learn More
TIGD1 Mouse anti-Human, Clone: OTI4G9, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF800156
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene belongs to the tigger subfamily of the pogo superfamily of DNA-mediated transposons in humans. These proteins are related to DNA transposons found in fungi and nematodes, and more distantly to the Tc1 and mariner transposases. They are also very similar to the major mammalian centromere protein B. The exact function of this gene is not known.Specifications
| TIGD1 | |
| Monoclonal | |
| Unconjugated | |
| TIGD1 | |
| EEYORE | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 200765 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4G9 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q96MW7 | |
| TIGD1 | |
| Human recombit protein fragment corresponding to amino acids 1-267 of human TIGD1 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.