missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TIGD1 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142098
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:16,000. Preliminary analysis showed no signal in Western Blot.
The protein encoded by this gene belongs to the tigger subfamily of the pogo superfamily of DNA-mediated transposons in humans. These proteins are related to DNA transposons found in fungi and nematodes, and more distantly to the Tc1 and mariner transposases. They are also very similar to the major mammalian centromere protein B. The exact function of this gene is not known.
Specifications
| TIGD1 | |
| Polyclonal | |
| Unconjugated | |
| TIGD1 | |
| EEYORE; TIGD1; tigger transposable element derived 1; tigger transposable element-derived protein 1 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 200765 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunocytochemistry | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q96MW7 | |
| TIGD1 | |
| Peptide with sequence C-PAKRVRLTEGSD. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction