missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIGD5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13644-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene belongs to the tigger subfamily of the pogo superfamily of DNA-mediated transposons in humans. These proteins are related to DNA transposons found in fungi and nematodes, and more distantly to the Tc1 and mariner transposases. They are also very similar to the major mammalian centromere protein B. The exact function of this gene is not known.Specifications
| TIGD5 | |
| Polyclonal | |
| Unconjugated | |
| TIGD5 | |
| TIGD5 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 105734, 300034, 84948 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.39 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| B1WC39, Q499M4, Q53EQ6 | |
| TIGD5 | |
| TIGD5 Fusion Protein Ag4590 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction