missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIGIT Antibody (2190A) [PerCP], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB72671C
This item is not returnable.
View return policy
Description
TIGIT Monoclonal antibody specifically detects TIGIT in Mouse samples. It is validated for Flow Cytometry, CyTOF
Specifications
| TIGIT | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry, CyTOF | |
| 2190A | |
| Flow Cytometry, CyTOF-ready | |
| T cell immunoreceptor with Ig and ITIM domains, TIGIT, VSIG9, VSTM3, WUCAM | |
| Mouse myeloma cell line NS0-derived recombinant mouse TIGIT, Met1-Thr143, Accession # P86176 | |
| 0.1 mL | |
| Cancer, Immunology, Virology Bacteria and Parasites | |
| 201633 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction