missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIGIT Antibody (2629A), Allophycocyanin/Cy7, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB10532APCCY7
This item is not returnable.
View return policy
Description
TIGIT Monoclonal antibody specifically detects TIGIT in Cynomolgus Monkey samples. It is validated for Flow Cytometry
Specifications
| TIGIT | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Cynomolgus Monkey | |
| Purified |
| Flow Cytometry | |
| 2629A | |
| Flow Cytometry | |
| T cell immunoreceptor with Ig and ITIM domains, TIGIT, VSIG9, VSTM3, WUCAM | |
| Human embryonic kidney cell HEK293-derived Cynomolgus monkey TIGIT, Met22-Pro142, Accession # XP_00554815 | |
| 0.1 mL | |
| Cancer, Immunology, Virology Bacteria and Parasites | |
| 201633 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction