missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIGIT Antibody (2629A) [Allophycocyanin], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB10532A
This item is not returnable.
View return policy
Description
TIGIT Monoclonal antibody specifically detects TIGIT in Cynomolgus Monkey samples. It is validated for Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF
Specifications
| TIGIT | |
| Monoclonal | |
| APC | |
| PBS | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Cynomolgus Monkey | |
| Purified |
| Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF | |
| 2629A | |
| Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-ready | |
| T cell immunoreceptor with Ig and ITIM domains, TIGIT, VSIG9, VSTM3, WUCAM | |
| Human embryonic kidney cell HEK293-derived Cynomolgus monkey TIGIT, Met22-Pro142, Accession # XP_00554815 | |
| 0.1 mL | |
| Cancer, Immunology, Virology Bacteria and Parasites | |
| 201633 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction