missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIGIT Monoclonal Antibody (GIGD7), Brilliant Violet™ 650, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen 416-9501-80
This item is not returnable.
View return policy
Description
Binds with high affinity to the poliovirus receptor (PVR) which causes increased secretion of IL10 and decreased secretion of IL12B and suppresses T cell activation by promoting the generation of mature immunoregulatory dendritic cells.
Specifications
| TIGIT | |
| Monoclonal | |
| 0.2 mg/mL | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| P86176 | |
| TIGIT | |
| mTIGIT-Fc treated with 4% PFA | |
| 25 μg | |
| Primary | |
| Mouse | |
| Liquid |
| Flow Cytometry | |
| GIGD7 | |
| Brilliant Violet 650 | |
| TIGIT | |
| ENSMUSG00000071552; T cell immunoreceptor with Ig and ITIM domains; T-cell immunoreceptor with Ig and ITIM domains; TIGIT; V-set and immunoglobulin domain containing 9; V-set and immunoglobulin domain-containing protein 9; V-set and transmembrane domain containing 3; V-set and transmembrane domain-containing protein 3; VSIG9; VSTM3; Washington University cell adhesion molecule; WUCAM | |
| Rat | |
| Affinity chromatography | |
| RUO | |
| 100043314 | |
| 4° C, store in dark, DO NOT FREEZE! | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction